1X4 Color™ B488 Dye Set
Supplier: AAT Bioquest
Warning
8005
76485-702EA 1142.29 USD
76485-702
1X4 Color™ B488 Dye Set
1X4 Color™ B488 Dye Set
1X4 Color™ B488 Dye Set provides a set of highly fluorescent dyes for multicolor applications.
  • all-in-one wavelength-excitable dye set
  • single laser excitation for multicolor detection
ADR: 5,II