Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*, AAT Bioquest
Supplier: AAT BIOQUEST INC
20130
76485-692EA 358.05 USD
76485-692
Cell Meter™ Live Cell Caspase 3/7 Imaging Kit *Green Fluorescence*, AAT Bioquest
Assays Cellular Assays
Cell Meter™ Live Cell No Wash Caspase 3/7 Imaging Kit uses our recently developed cell-permeable fluorogenic caspase substrate, ApoSight™ Green Caspase 3/7, the first fluorogenic probe, for the direct detection of caspase activities in live cells.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.