ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole), AAT Bioquest
Supplier: AAT BIOQUEST INC
17207
76485-678EA 237.74 USD
76485-678
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole), AAT Bioquest
Nucleic Acid Reagents Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.