iFluor® 555 fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1016
76485-612EA 237.74 USD
76485-612
iFluor® 555 fluorescent probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84f469790814f30f31be75e2c747f6ce8d6f4e8d&vid=84f469790814f30f31be75e2c747f6ce8d6f4e8d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=290cb3fd712f5de3543c92f8ffa93ca52e660610&vid=f5230cc3d1ca5b0da50be5f7e4b0f8ab9b2e641c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR