iFluor® 488 fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1014
76485-610EA 237.74 USD
76485-610
iFluor® 488 fluorescent probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4380bb0745fd8aa62e6163b19ee8a3a4a704e706&vid=4380bb0745fd8aa62e6163b19ee8a3a4a704e706&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b05631f7cfb5205b7e74a6bf57e81d14ca6d19a&vid=2f646a24d01809daf55203df7f58a01685160ccb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR