Blue 580 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: Blue 580 SE

1178
76485-580EA 358.05 USD
76485-580
Blue 580 succinimidyl ester, mFluor™ fluorescent probe
Blue 580 succinimidyl ester

Advances in spectral flow cytometers have expanded applications and capabilities beyond conventional flow cytometry.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 832.98 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5380e19fb342c73e49f1e8abc5016db455170557&vid=5380e19fb342c73e49f1e8abc5016db455170557&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc52b98616cf2fd11cbaa8bce7cfff34e8cf7e13&vid=c78607011283d605ce019c9dd1b59d73fe689486&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR