CDPI3 Azide DNA binding
Supplier: AAT BIOQUEST INC
Warning
6909
76485-520EA 237.74 USD
76485-520
CDPI3 Azide DNA binding
CDPI3 Azide
Minor Groove Binder (MGB) molecules selectively bind to the minor groove of a DNA molecule, the shallow furrow in the DNA helix.
  • Stable binding
  • Hybridization probe precursor
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9284f3725f74d891236fd1d3c1f3cf52838eb6df&vid=9284f3725f74d891236fd1d3c1f3cf52838eb6df&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddcf500eb9ffcb47c81090fd1f0381335b91dc63&vid=c0bc2235eb4fb74191479936776dc91bbc424eb8&prid=web&clid=SAPDAM