CDPI3 Maleimide DNA binding
Supplier: AAT BIOQUEST INC
Warning
6908
76485-518EA 237.74 USD
76485-518
CDPI3 Maleimide DNA binding
CDPI3 Maleimide
Minor Groove Binder (MGB) molecules selectively bind to the minor groove of a DNA molecule, the shallow furrow in the DNA helix.
  • Stable binding
  • Hybridization probe precursor
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39dd16a8488c1712f7cfb79d34f3793240b2448d&vid=de9ea4a9475d62e4a6f2ec5df3de264fddfae273&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d5a0dc753afa03afbdcdc3b78b7320c1c176be5&vid=7d5a0dc753afa03afbdcdc3b78b7320c1c176be5&prid=web&clid=SAPDAM