Tetracysteine FLASH Reagent *2 mM DMSO Solution*, AAT Bioquest
Supplier: AAT BIOQUEST INC
22330
76485-500EA 462.11 USD
76485-500
Tetracysteine FLASH Reagent *2 mM DMSO Solution*, AAT Bioquest
Assays Protein Assays
FLASH is a fluorescein derivative, modified to contain two arsenic atoms at a set distance from each other.
  • Fluorescein derivative
  • High affinity for thiols
  • Multiple applications
Caution: For research use only.