Estradiol succinimidyl ester

Supplier: AAT Bioquest
Warning

50545
76485-410EA 117.59 USD
76485-410
Estradiol succinimidyl ester
Estradiol succinimidyl ester

Estradiol is an estrogen steroid hormone and the major female sex hormone.


  • assay development building block
  • multiple research applications
  • in-house quality control

Storage Temperature: Freezer
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR