Biotin-PEG4-tyramide
Supplier: AAT BIOQUEST INC
45352
76485-386EA 117.59 USD
76485-386
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide is a reagent that has been used for tyramide signal amplification (TSA) via catalyzed reporter deposition (CARD).
  • increased fluorescence signal
  • suitable for low-abundance targets
  • compatible with all major fluorescence platforms
Caution: For research use only