iFluor® 594, Tyramide
Supplier: AAT BIOQUEST INC
45107
76485-382EA 358.05 USD
76485-382
iFluor® 594, Tyramide
iFluor®
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • increased fluorescence signal
  • suitable for low-abundance targets
  • compatible with all major fluorescence platforms
Caution: For research use only