Concanavalin A, XFD488
Supplier: AAT Bioquest
25570
76485-364EA 117.59 USD
76485-364
Concanavalin A, XFD488
Proteins and Peptides
XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).
  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
Caution: For research use only.