Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Supplier: AAT Bioquest
25550
76485-360EA 117.59 USD
76485-360
Native Human Wheat Germ Agglutinin (from Eukaryotic cells), iFluor® 594
Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.
  • Glycoconjugate binding
  • Very bright signal
  • Multiple applications
Caution: For research use only.