BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking

Supplier: AAT BIOQUEST INC
Warning

Synonyms: 2′,7′-Bis(2-carboxyethyl)-5(6′)-carboxyfluorescein acetoxymethyl ester, Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-2',7'-dipropanoic acid, BCECF AM

21200
76485-216EA 358.05 USD
76485-216
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Cell permeable
  • Improved cell retention
  • In-house quality testing


Caution: For research use only

Formula: C₄₂H₄₀O₂₁
MDL Number: MFCD00036969
CAS Number: 117464-70-7
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR