XFD568 NHS Ester fluorescent probe

Supplier: AAT Bioquest
Warning

1823
76485-210EA 237.74 USD
76485-210
XFD568 NHS Ester fluorescent probe
XFD568 NHS Ester

XFD568 has the same chemical structure of Alexa Fluor® 568 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

CAS Number: 247145-38-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR