Dextran, M.W. 150,000 FITC labelled

Supplier: AAT BIOQUEST INC

21704
76485-176EA 177.52 USD
76485-176
Dextran, M.W. 150,000 FITC labelled
Dextran

FITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.


  • Well-retained in cells
  • 150,000 mw
  • Multiple applications


From Eukaryotic cells


Caution: For research use only.

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR