DiYO™-1 [Equivalent To YOYO®-1], AAT Bioquest
Supplier: AAT Bioquest
17579
76485-130EA 311.76 USD
76485-130
DiYO™-1 [Equivalent To YOYO®-1], AAT Bioquest
Assays Cellular Assays
DiYO™-1 is chemically equivalent to YOYO®-1 (YOYO® is the trademark of Invitrogen).
  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
Caution: For Research Use Only