CellPaint™ TSP membrane stain, AAT Bioquest
Supplier: AAT Bioquest
22700
76485-108EA 249.58 USD
76485-108
CellPaint™ TSP membrane stain, AAT Bioquest
Assays Cellular Assays
TSP is a styrylpyridine-based fluorescent membrane probe suitable for imaging plasma membranes in living cells and tissues.
  • Low Cytotoxicity
  • High Target Affinity
  • Bright Fluorescence
Caution: For Research Use Only