UV460 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

Synonyms: UV460 SE

1136
76485-074EA 249.58 USD
76485-074
UV460 succinimidyl ester, mFluor™ fluorescent probe
UV460 succinimidyl ester

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 367.26 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0a640f5199d58d068fd2acb68d053d8acbc80b7&vid=a0a640f5199d58d068fd2acb68d053d8acbc80b7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR