CytoCite™ Sample Tubes (500 Tubes), AAT Bioquest
Supplier: AAT Bioquest
CCT100
76484-962EA 104.68 USD
76484-962
CytoCite™ Sample Tubes (500 Tubes), AAT Bioquest
Tubes Test Tubes
Cytocite™ sample tubes are 500 µl thin-walled polypropylene tubes validated for use with Cytocite or Qubit Fluorometer.
  • Excellent quality fluorometer sample tube
  • Fully validated