PSA™ iFluor® 594 Imaging Kit with Goat Anti-Mouse IgG
Supplier: AAT Bioquest
45280
76484-930EA 626.51 USD
76484-930
PSA™ iFluor® 594 Imaging Kit with Goat Anti-Mouse IgG
Protein Linking and Labeling Systems
iFluor® 594 PSA kit is a much superior replacement for Alexa Fluor 594 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.