Ifluor 350/Trade Ps 45250, Power Styramide/trade; Signal Amplification (PSA/trade;) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Supplier: AAT Bioquest
Ifluor 350/Trade Ps 45250, Power Styramide/trade; Signal Amplification (PSA/trade;) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.