iFluor® 594, Styramide *Superior Replacement for Alexa Fluor 594 tyramide*
Supplier: AAT BIOQUEST INC
45035
76484-898EA 375.27 USD
76484-898
iFluor® 594, Styramide *Superior Replacement for Alexa Fluor 594 tyramide*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
  • 10-50x higher fluorescence
  • improved precision and sensitivity
  • more efficient use of primary antibody
  • enhanced water solubility
  • more photostable
Caution: For research use only