Bis-Arg-rhodamine 110
Supplier: AAT Bioquest
Warning
13481
76484-864EA 375.27 USD
76484-864
Bis-Arg-rhodamine 110
Bis-Arg-rhodamine 110
This Bis Arg-rhodamine 110 is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of human alpha-thrombin.
  • alpha-thrombin substrate
  • bright signal
  • multiple applications
ADR: 5,II