Red 780 acid, mFluor™ fluorescent probe
Supplier: AAT Bioquest
1147
76484-842EA 375.27 USD
76484-842
Red 780 acid, mFluor™ fluorescent probe
mFluor™ Red 780 dyes are an excellent alternative to APC-Alexa Fluor® 750 tandems since they have the spectral properties equivalent to those of APC-Alexa Fluor® 750 conjugates.
  • pH-insensitive
  • High photostability
  • Brighter signal