ICG azide fluorescent probe
Supplier: AAT BIOQUEST INC
Warning
985
76484-808EA 249.58 USD
76484-808
ICG azide fluorescent probe
ICG azide
Indocyanine green (ICG) is a tricarbocyanine-type dye with NIR-absorbing properties (peak absorption around 780 nm) and emission maximum at ~800 nm.
  • Low autofluorescence
  • NIR fluorescence
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0cf9d22d7d642c923f856cd0d2e4e2b9ba6ae10&vid=3d8233a1e0bdf14b8f6114eeebb56a5381256a44&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d28b81b6a020be3fceaa920c8f6058f08e50cb47&vid=d28b81b6a020be3fceaa920c8f6058f08e50cb47&prid=web&clid=SAPDAM