Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

2274
76484-774EA 500.94 USD
76484-774
Tide Fluor™ 3 phosphoramidite, Tide Fluor™ fluorescent probe
Tide Fluor™ 3 phosphoramidite

Tide Fluor™ 3 (TF3) phosphoramidite is a high-performance alternative to Cy3 phosphoramidite for the in-synthesis labeling of oligonucleotides.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf867f76a63d50a6d309def81af85d44fd90ed69&vid=bf867f76a63d50a6d309def81af85d44fd90ed69&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6af8468f1b40f8053f306c86a590a742fbe08f86&vid=0ddbdcd4e887259db042a0fe98bb1e3ab4e14b6f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR