MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit, AAT Bioquest
Supplier: AAT Bioquest
22405
76484-738EA 375.27 USD
76484-738
MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit, AAT Bioquest
Assays Immunological Assays
The CTC Flow Cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial health and vitality as a function of the respiratory activity.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for flow cytometry
Caution: For research use only.