XFD532 acid fluorescent probe

Supplier: AAT BIOQUEST INC
Warning

1795
76484-722EA 500.94 USD
76484-722
XFD532 acid fluorescent probe
XFD532 acid

XFD532 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 532 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR