m-PEG24-Mal for biomolecular labeling

Supplier: AAT Bioquest
Warning

4525
76484-720EA 124.31 USD
76484-720
m-PEG24-Mal for biomolecular labeling
m-PEG24-Mal

MM(PEG)24 is a methyl-terminated, polyethylene glycol compound (24 PEG units) activated with a maleimide group for covalent pegylation of sulfhydryls on biomolecules or assay surfaces.


  • Improved crosslinking efficiency
  • High water solubility

Formula: C₅₆H₁₀₆N₂O₂₇
MDL Number: MFCD11041152
CAS Number: 88504-24-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f26c4a63742b9ac2f4feaa5f8b0b5fab2ac555c&vid=8f26c4a63742b9ac2f4feaa5f8b0b5fab2ac555c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a67947e77909c3e417badc56cd989beebbddf1&vid=b2f4a50e696649eb46ef4b12e955a7e241cecc8e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR