Xfd488 Phalloidin 23153, XFD488 has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher). XFD488 phalloidin conjugate is chemically equivalent to Alexa Fluor® 488 phalloidin.
Supplier: AAT Bioquest
Xfd488 Phalloidin 23153, XFD488 has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher). XFD488 phalloidin conjugate is chemically equivalent to Alexa Fluor® 488 phalloidin.