Fmoc-Lys(biotin)-OH for biomolecular labeling

Supplier: AAT Bioquest
Warning

5020
76484-664EA 124.31 USD
76484-664
Fmoc-Lys(biotin)-OH for biomolecular labeling
Fmoc-Lys(biotin)-OH

FMOC-Lys(Biotin)-OH is a lysine derivative for the preparation of biotin-labeled peptides by FMOC-SPPS.


  • FRET building block
  • Multiple applications

Formula: C₃₁H₃₈N₄O₆S
MDL Number: MFCD00270741
CAS Number: 146987-10-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d24c58670a784b912f180d48d985c58afe070f33&vid=d24c58670a784b912f180d48d985c58afe070f33&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03eff458becc5e4870c3367475aec851c9ffa876&vid=642b97f109d52587e894a9396033344e18ce0171&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR