Cal-670™ potassium salt
Supplier: AAT Bioquest
Warning
20455
76484-640EA 510.51 USD
76484-640
Cal-670™ potassium salt
Cal-670™ potassium salt
Cal-670™ is a near infrared (NIR) calcium indicator with maximum emission at ~675 nm.
  • Stable for 12 months
  • Kd ~853 nM
Calcium measurement is critical for numerous biological investigations.
Caution: For Research Use Only
ADR: 5,II