Violet 500 succinimidyl ester, mFluor™ fluorescent probe

Supplier: AAT Bioquest
Warning

1149
76484-542EA 249.58 USD
76484-542
Violet 500 succinimidyl ester, mFluor™ fluorescent probe
Violet 500 succinimidyl ester

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b101362a45677e502c76507a47f94a78b204579e&vid=b101362a45677e502c76507a47f94a78b204579e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR