XFD488 C5 Maleimide fluorescent probe
Supplier: AAT Bioquest
1878
76484-516EA 249.58 USD
76484-516
XFD488 C5 Maleimide fluorescent probe
XFD488 C5 Maleimide is the same molecule to the Alexa Fluor® 488 C5 Maleimide (Alexa Fluor® is the trademark of ThermoFisher).
  • pH-insensitive
  • High photostability
  • Brighter signal