iFluor® 594, Maleimide

Supplier: AAT BIOQUEST INC
Warning

1064
76484-424EA 249.58 USD
76484-424
iFluor® 594, Maleimide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e8cdb47b69d09c298c5b2b4f70e9845950069e8&vid=5e8cdb47b69d09c298c5b2b4f70e9845950069e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR