6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*, AAT Bioquest
Supplier: AAT Bioquest
398
76484-268EA 375.27 USD
76484-268
6-ROXtra™ Fluorescence Reference Solution *25 µM for PCR Reactions*, AAT Bioquest
Assays qPCR Assays
6-ROX is predominately used as a reference dye for performing PCR detections.
  • Multiple Bioconjugation Applications
  • 95% Purity
Caution: For Research Use Only