Hrp-Labeled Goat Ant 16793 1Mg, The enzyme labels can be visualized by means of enzyme histochemical methods via chromogenic (producing color) reactions, or via fluorogenic (producing fluorescence) reactions. The most used enzyme is horse radish peroxidase (HRP), Size: 1mg
Supplier: AAT Bioquest
Hrp-Labeled Goat Ant 16793 1Mg, The enzyme labels can be visualized by means of enzyme histochemical methods via chromogenic (producing color) reactions, or via fluorogenic (producing fluorescence) reactions. The most used enzyme is horse radish peroxidase (HRP), Size: 1mg