Fluo-5N AM Cell permeant
Supplier: AAT BIOQUEST INC
Warning
20566
76484-242EA 249.58 USD
76484-242
Fluo-5N AM Cell permeant
Fluo-5N AM
Fluo-5N is an analog of Fluo-4 with lower calcium-binding affinity (Kd = ~90 uM), making it suitable for detecting intracellular calcium levels in the range of 1 µM to 1 mM that would saturate the response of Fluo-4.
  • Calcium-dependent fluorescence
  • Steady signal
  • Cell permeant
Caution: For research use only
ADR: 5,II