OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
Supplier: AAT Bioquest
Warning
20506
76484-234EA 249.58 USD
76484-234
OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
OG488 BAPTA-1 hexapotassium salt
OG488 BAPTA -1 is the same molecule of Oregon Green 488 BAPTA-1.
  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant
Caution: For research use only
ADR: 5,II