iFluor® 820, Maleimide
Supplier: AAT BIOQUEST INC
Warning
1398
76484-190EA 375.27 USD
76484-190
iFluor® 820, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249d826077a1c532eb54845bda2a2378bd9f0f95&vid=1e4888ab06fe2e4cb613188f82b355d01bc32747&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=968790478971c30b5c8a9987df0f4afe2ef2baec&vid=968790478971c30b5c8a9987df0f4afe2ef2baec&prid=web&clid=SAPDAM