CLF [Cholyl-Lys-Fluorescein]

Supplier: AAT Bioquest
Warning

36701
76483-998EA 375.27 USD
76483-998
CLF [Cholyl-Lys-Fluorescein]
CLF [Cholyl-Lys-Fluorescein]

CLF is a fluorescein-labeled bile acid analog that closely parallels the cellular binding and uptake properties of cholic acid derivatives.


  • well-cited
  • stable signal
  • multiple applications

Storage Temperature: Freezer
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR