Screen Quest™ 10X Cell Staining Buffer with Phenol Red Plus™, AAT Bioquest
Supplier: AAT Bioquest
36300
76483-920EA 124.31 USD
76483-920
Screen Quest™ 10X Cell Staining Buffer with Phenol Red Plus™, AAT Bioquest
Ready-to-use buffer that is optimized for fluorescence cell imaing.
  • Improves fluorescent signal
  • Ready to use solution
  • In-house quality control
Caution: For research use only.