MycoLight™ Green JJ98 *5 mM in DMSO*, AAT Bioquest
Supplier: AAT BIOQUEST INC
24000
76483-902EA 249.58 USD
76483-902
MycoLight™ Green JJ98 *5 mM in DMSO*, AAT Bioquest
Assays Cellular Assays
MycoLight™ Green JJ98 is a green-fluorescent nuclear and chromosome stain that is permeant to both prokaryotic and eukaryotic cell membranes.
  • Cell-Permeable
  • High DNA Affinity
  • Bright Fluorescence
Caution: For Research Use Only