Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor® 514
Supplier: AAT Bioquest
23116
76483-874EA 188.87 USD
76483-874
Native Amanita phalloides Phalloidin (from Eukaryotic cells), iFluor® 514
Proteins and Peptides
This yellow fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 514-labeled phalloidin) selectively binds to F-actins.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.