Congo red ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST INC
Warning

23056
76483-854EA 96.26 USD
76483-854
Congo red ≥95%, Ultra Pure Grade
Congo Red

Congo red (CR) is the sodium salt of benzidinediazo-bis-1-naphtylamine-4-sulfonic acid.

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MDL Number: MFCD00004028
CAS Number: 573-58-0
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR