Cell Navigator® Cell Plasma Membrane Staining Kit *Orange Fluorescence*, AAT Bioquest
Supplier: AAT BIOQUEST INC
22680
76483-706EA 249.58 USD
76483-706
Cell Navigator® Cell Plasma Membrane Staining Kit *Orange Fluorescence*, AAT Bioquest
Assays Cellular Assays
The Cell Navigator® Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins.
  • Signal persists through fixation
  • Stable labeling regardless of cell type
  • Multiple applications
Caution: For research use only.