MitoLite™ Orange EX405, AAT Bioquest
Supplier: AAT BIOQUEST INC
22679
76483-704EA
124.31
USD
76483-704
MitoLite™ Orange EX405, AAT Bioquest
Assays
Cellular Assays
MitoLite™ Orange 405 is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
Caution: For Research Use Only