MitoLite™ Orange FX570, AAT Bioquest
Supplier: AAT Bioquest
22676
76483-698EA
124.31
USD
76483-698
MitoLite™ Orange FX570, AAT Bioquest
Assays
Cellular Assays
MitoLite™ Orange is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
Caution: For Research Use Only